missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2C (phospho S396), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MEF2C.
Brand: Abnova PAB29248.100uL
This item is not returnable.
View return policy
Specifications
| MEF2C | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MEF2C. | |
| In PBS, pH 7.4 (without Mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) | |
| MEF2C | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S396 of human MEF2C. | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of MEF2C only when phosphorylated at serine 396. | |
| Store at -20°C. Avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user. | |
| MEF2C | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 4208 | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction