missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED19 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MED19.
Brand: Abnova PAB23597.100uL
This item is not returnable.
View return policy
Description
MED19 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II (Sato et al., 2003 [PubMed 12584197]).[supplied by OMIM
Sequence: GSHDNSSLRSLIEKPPILSSSFNPITGTMLAGFRLHTGPLPEQCRLMHIQPPKKKNKHKHKQSRTQDPVSpecifications
| MED19 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MED19. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DT2P1G7/LCMR1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 219541 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| MED19 | |
| MED19 | |
| Recombinant protein corresponding to amino acids of human MED19. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction