Learn More
MED12 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MED12.
Brand: Abnova PAB28711.100uL
Description
The initiation of transcription is controlled in part by a large protein assembly known as the preinitiation complex. A component of this preinitiation complex is a 1.2mDa protein aggregate called Mediator. This Mediator component binds with a CDK8 subcomplex which contains the protein encoded by this gene, mediator complex subunit 12 (MED12), along with MED13, CDK8 kinase, and cyclin C. The CDK8 subcomplex modulates Mediator-polymerase II interactions and thereby regulates transcription initiation and reinitation rates. The MED12 protein is essential for activating CDK8 kinase. Defects in this gene cause X-linked Opitz-Kaveggia syndrome, also known as FG syndrome, and Lujan-Fryns syndrome. [provided by RefSeq
Sequence: LPTTMTGVMGLEPSSYKTSVYRQQQPAVPQGQRLRQQLQAKIQSQGMLGQSSVHQMTPSSSYGLQTSQGYTPYVSHVGLQQHTGPAGTMVPPSYSSQPYQSTHPSTNPTLVDPTRHLQQRPSGYVHQQAPTYGHGLTSSpecifications
| MED12 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MED12. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| CAGH45/FGS1/HOPA/KIAA0192/OKS/OPA1/TNRC11/TRAP230 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9968 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| MED12 | |
| MED12 | |
| Recombinant protein corresponding to amino acids of human MED12. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only