missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Med, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Med.
Brand: Abnova PAB8968.100ug
This item is not returnable.
View return policy
Description
Sequence: YGYPQGPSEYVGDANPSpecifications
| Med | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Med. | |
| In buffer containing 0.02% sodium azide | |
| CG1775/E(zen)3/anon-EST:Posey121/dSmad4/l(3)11m-254/l(3)12m-137/l(3)SG36/l(3)SG70/l(3)XIIm137/medea | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Fruitfly | |
| Liquid |
| Dot Blot, ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:20000-1:50000) Dot Blot (1:20000-1:50000) Western Blot (1:500-1:750) The optimal working dilution should be determined by the end user. | |
| Med | |
| Med | |
| A synthetic peptide corresponding to amino acids 194-209 of fruit fly Med. | |
| RUO | |
| 43725 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction