Learn More
ME2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ME2.
Brand: Abnova PAB17785.100ug
Description
This gene encodes a mitochondrial NAD-dependent malic enzyme, a homotetrameric protein, that catalyzes the oxidative decarboxylation of malate to pyruvate. It had previously been weakly linked to a syndrome known as Friedreich ataxia that has since been shown to be the result of mutation in a completely different gene. [provided by RefSeq
Specifications
| ME2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ME2. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| ME2 | |
| A synthetic peptide corresponding to internal of human ME2. | |
| RUO | |
| 4200 | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| ME2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total ME2 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only