Learn More
MDM2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MDM2.
Brand: Abnova PAB27165.100uL
Description
This gene is a target gene of the transcription factor tumor protein p53. The encoded protein is a nuclear phosphoprotein that binds and inhibits transactivation by tumor protein p53, as part of an autoregulatory negative feedback loop. Overexpression of this gene can result in excessive inactivation of tumor protein p53, diminishing its tumor suppressor function. This protein has E3 ubiquitin ligase activity, which targets tumor protein p53 for proteasomal degradation. This protein also affects the cell cycle, apoptosis, and tumorigenesis through interactions with other proteins, including retinoblastoma 1 and ribosomal protein L5. More than 40 different alternatively spliced transcript variants have been isolated from both tumor and normal tissues. [provided by RefSeq
Specifications
| MDM2 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| MDM2 | |
| MDM2 | |
| A synthetic peptide corresponding to human MDM2. | |
| 100 μL | |
| Primary | |
| MDM2 polyclonal antibody detects endogenous levels of MDM2 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of MDM2. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| HDMX/MGC71221/hdm2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 4193 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only