Learn More
MDC1 (phospho S513), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MDC1.
Brand: Abnova PAB29247.100uL
Description
The protein encoded by this gene contains an N-terminal forkhead domain, two BRCA1 C-terminal (BRCT) motifs and a central domain with 13 repetitions of an approximately 41-amino acid sequence. The encoded protein is required to activate the intra-S phase and G2/M phase cell cycle checkpoints in response to DNA damage. This nuclear protein interacts with phosphorylated histone H2AX near sites of DNA double-strand breaks through its BRCT motifs, and facilitates recruitment of the ATM kinase and meiotic recombination 11 protein complex to DNA damage foci. [provided by RefSeq
Specifications
| MDC1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MDC1. | |
| In PBS, pH 7.4 (without Mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) | |
| DKFZp781A0122/KIAA0170/MGC166888/NFBD1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 9656 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| MDC1 | |
| MDC1 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S513 of human MDC1. | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of MDC1 only when phosphorylated at serine 513. | |
| Store at -20°C. Avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only