Learn More
MCM5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MCM5.
Brand: Abnova PAB27075.100uL
Description
The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation. [provided by RefSeq
Specifications
| MCM5 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| MCM5 | |
| MCM5 | |
| A synthetic peptide corresponding to human MCM5. | |
| 100 μL | |
| Primary | |
| MCM5 polyclonal antibody detects endogenous levels of MCM5 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of MCM5. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| CDC46/MGC5315/P1-CDC46 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 4174 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only