Learn More
MCHR1 Rabbit anti-Bovine, Dog, Human, Monkey, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MCHR1.
Brand: Abnova PAB16225.50ug
Description
The protein encoded by this gene, a member of the G protein-coupled receptor family 1, is an integral plasma membrane protein which binds melanin-concentrating hormone. The encoded protein can inhibit cAMP accumulation and stimulate intracellular calcium flux, and is probably involved in the neuronal regulation of food consumption. Although structurally similar to somatostatin receptors, this protein does not seem to bind somatostatin. [provided by RefSeq
Specifications
| MCHR1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MCHR1. | |
| In PBS (0.09% sodium azide) | |
| GPR24/MCH1R/MGC32129/SLC1 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 2847 | |
| Bovine, Canine, Human, Monkey, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| MCHR1 | |
| MCHR1 | |
| A synthetic peptide (conjugated with KLH) corresponding to human MCHR1. | |
| 50 μg | |
| Primary | |
| C-terminal domain of human. Predicted crossreactivity with mouse and rat due to sequence similarity. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only