missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP (Maltose Binding Protein) Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MBP (Maltose Binding Protein).
Brand: Abnova PAB10344.1mg
This item is not returnable.
View return policy
Specifications
| MBP (Maltose Binding Protein) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MBP (Maltose Binding Protein). | |
| Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Recombinant protein corresponding to MBP (Maltose Binding Protein). | |
| RUO | |
| This polyclonal anti-MBP epitope tag antibody detects over-expressed proteins containing the MBP epitope tag. | |
| Lyophilized |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:4000-1:16000) Western Blot (1:3500) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 1 mg | |
| Primary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction