Learn More
MBD4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MBD4.
Brand: Abnova PAB20104.100uL
Description
DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. Human proteins MECP2, MBD1, MBD2, MBD3, and MBD4 comprise a family of nuclear proteins related by the presence in each of a methyl-CpG binding domain (MBD). Each of these proteins, with the exception of MBD3, is capable of binding specifically to methylated DNA. MBD4 may function to mediate the biological consequences of the methylation signal. In addition, MBD4 has protein sequence similarity to bacterial DNA repair enzymes and thus may have some function in DNA repair. Further, MBD4 gene mutations are detected in tumors with primary microsatellite-instability (MSI), a form of genomic instability associated with defective DNA mismatch repair, and MBD4 gene meets 4 of 5 criteria of a bona fide MIS target gene. [provided by RefSeq
Sequence: LQNQSNNSNWNLRTRSKCKKDVFMPPSSSSELQESRGLSNFTSTHLLLKEDEGVDDVNFRKVRKPKGKVTILKGIPIKKTKKGCRKSCSGFVQSDSKRESVCNKADAESEPVAQKSQLDRTVCISDAGACGETLSVTSEENSpecifications
| MBD4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MBD4. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| MED1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8930 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| MBD4 | |
| MBD4 | |
| Recombinant protein corresponding to amino acids of human MBD4. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only