Learn More
MASP1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MASP1
Brand: Abnova PAB28463.100uL
Description
The Ra-reactive factor (RARF) is a complement-dependent bactericidal factor that binds to the Ra and R2 polysaccharides expressed by certain enterobacteria. Alternate splicing of this gene results in multiple transcript variants encoding two RARF components that are involved in the mannan-binding lectin (MBL) pathway of complement activation. Two of the isoforms are cleaved into two chains which form a heterodimer linked by a disulfide bond. The encoded proteins are members of the trypsin family of peptidases. [provided by RefSeq
Sequence: PCPYDYIKIKVGPKVLGPFCGEKAPEPISTQSHSVLILFHSDNSGENRGWRLSYRAAGNECPELQPPVHGKIEPSQAKYFFKDQVLVSCDTGYKVLKDNVEMDTFQIECLKDGTWSNKIPTSpecifications
| MASP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MASP1 | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| CRARF/CRARF1/DKFZp686I01199/FLJ26383/MASP/MGC126283/MGC126284/PRSS5/RaRF | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5648 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry(1:20-1:50) The optimal working dilution should be determined by the end user. | |
| MASP1 | |
| MASP1 | |
| Recombinant protein corresponding to amino acids of human MASP1 | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only