missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK9/MAPK8 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAPK9/MAPK8.
Brand: Abnova PAB18484.100ug
This item is not returnable.
View return policy
Specifications
| MAPK9/MAPK8 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| JNK-55/JNK2/JNK2A/JNK2ALPHA/JNK2B/JNK2BETA/PRKM9/SAPK/p54a/p54aSAPK | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 5601/5599 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of MAPK9/MAPK8. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| MAPK9 | |
| A synthetic peptide corresponding to residues surrounding T183 of human MAPK9/MAPK8. | |
| 100 μg | |
| Primary | |
| This antibody is specific to MAPK9/MAPK8. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction