missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK9/MAPK10 (phospho T183), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAPK9/MAPK10.
Brand: Abnova PAB25838.100ug
This item is not returnable.
View return policy
Description
Sequence: M-M-Tp-P-YSpecifications
| MAPK9/MAPK10 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| JNK-55/JNK2/JNK2A/JNK2ALPHA/JNK2B/JNK2BETA/PRKM9/SAPK/p54a/p54aSAPK | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5601/5602 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAPK9/MAPK10. | |
| In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| MAPK9 | |
| Synthetic phosphopeptide corresponding to residues surrounding T183 of human MAPK9/MAPK10. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction