Learn More
MAPK7 (phospho S731/T733), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAPK7.
Brand: Abnova PAB15919.100ug
Description
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This kinase is specifically activated by mitogen-activated protein kinase kinase 5 (MAP2K5/MEK5). It is involved in the downstream signaling processes of various receptor molecules including receptor type kinases, and G protein-coupled receptors. In response to extracelluar signals, this kinase translocates to cell nucleus, where it regulates gene expression by phosphorylating, and activating different transcription factors. Four alternatively spliced transcript variants of this gene encoding two distinct isoforms have been reported. [provided by RefSeq
Specifications
| MAPK7 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAPK7. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| BMK1/ERK4/ERK5/PRKM7 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| MAPK7 | |
| MAPK7 | |
| Synthetic phosphopeptide corresponding to residues surrounding S731/T733 of human MAPK7. | |
| RUO | |
| 5598 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only