Learn More
MAP3K1 (phospho T1400), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MAP3K1.
Brand: Abnova PAB29207.100uL
Description
MAP3K, or MEK kinase, is a serine/threonine kinase that occupies a pivotal role in a network of phosphorylating enzymes integrating cellular responses to a number of mitogenic and metabolic stimuli, including insulin (MIM 176730) and many growth factors.[supplied by OMIM
Spezifikation
| MAP3K1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MAP3K1. | |
| In PBS, pH 7.4 (without Mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) | |
| MAPKKK1/MEKK/MEKK1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 4214 | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user. | |
| MAP3K1 | |
| MAP3K1 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T1400 of human MAP3K1. | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of MAP3K1 only when phosphorylated at threonine 1400. | |
| Store at -20°C. Avoid repeated freezing and thawing. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only