Learn More
MAP3K1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAP3K1.
Brand: Abnova PAB2272.400uL
Description
MAP3K, or MEK kinase, is a serine/threonine kinase that occupies a pivotal role in a network of phosphorylating enzymes integrating cellular responses to a number of mitogenic and metabolic stimuli, including insulin (MIM 176730) and many growth factors.[supplied by OMIM
Specifications
| MAP3K1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MAP3K1. | |
| In PBS (0.09% sodium azide) | |
| MAPKKK1/MEKK/MEKK1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 4214 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| MAP3K1 | |
| MAP3K1 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human MAP3K1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only