missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2K1/MAP2K2 Rabbit anti-Chicken, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAP2K1/MAP2K2.
Brand: Abnova PAB12707.100ug
This item is not returnable.
View return policy
Specifications
| MAP2K1/MAP2K2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MAP2K1/MAP2K2. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| MAP2K1 | |
| A synthetic peptide corresponding to residues surrounding S218 and S222 of human MAP2K1/MAP2K2. | |
| RUO | |
| 5604/5605 | |
| Chicken, Human, Mouse, Rat | |
| Liquid |
| Dot Blot, ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| MAPKK1/MEK1/MKK1/PRKMK1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody recognizes ∼44, 45 KDa of human MAP2K1 without phosphorylation sites of Ser 218/222 or MAP2K2 (pS222/226). It does not cross react to phospho MAP2K1/MAP2K2. | |
| Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction