missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2K1/MAP2K2 (phospho S218/S222), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP2K1/MAP2K2.
Brand: Abnova PAB9600.100uL
This item is not returnable.
View return policy
Specifications
| MAP2K1/MAP2K2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP2K1/MAP2K2. | |
| In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) | |
| MAP2K1 | |
| Synthetic phosphopeptide corresponding to residues surrounding S218/S222 of human MAP2K1/MAP2K2 . | |
| 100 μL | |
| Primary | |
| Bovine, Canine, Chicken, Frog, Human, Mouse, Primate, Rat, Zebrafish | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| MAPKK1/MEK1/MKK1/PRKMK1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 5604/5605 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction