Learn More
MAP1LC3B Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAP1LC3B.
Brand: Abnova PAB0736.100uL
Description
The product of this gene is a subunit of neuronal microtubule-associated MAP1A and MAP1B proteins, which are involved in microtubule assembly and important for neurogenesis. Studies on the rat homolog implicate a role for this gene in autophagy, a process that involves the bulk degradation of cytoplasmic component. [provided by RefSeq
Specifications
| MAP1LC3B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MAP1LC3B. | |
| Lyophilized from PBS | |
| LC3B/MAP1A/1BLC3 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Lyophilized |
| Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:100-1:1000) Immunofluorescence (1:100-1:1000) Western Blot (1:100-1:1000) The optimal working dilution should be determined by the end user. | |
| MAP1LC3B | |
| MAP1LC3B | |
| A synthetic peptide (conjugated with Blue Carrier Protein) corresponding to N-terminus of human MAP1LC3B. | |
| RUO | |
| 81631 | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only