missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEE1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MAGEE1.
Brand: Abnova PAB28639.100uL
This item is not returnable.
View return policy
Description
Sequence: STLFSSSASVDRNPSKCSLVLPSPRVTKASVDSDSEGPKGAEGPIEFEVLRDCESPNSISIMGLNTSRVAITLKPQDPMEQNVAELLQFLLVKDQSKYPIRESpecifications
| MAGEE1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MAGEE1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| DAMAGE/HCA1/KIAA1587 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 57692 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Immunohistochemistry (Formalin/Paraffin) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| MAGEE1 | |
| MAGEE1 | |
| Recombinant protein corresponding to amino acids of human MAGEE1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur