Learn More
MAGEA2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAGEA2.
Brand: Abnova PAB4744.400uL
Description
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. This gene has two identical copies at different loci. Alternatively spliced transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq
Specifications
| MAGEA2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MAGEA2. | |
| In PBS (0.09% sodium azide) | |
| MAGE2/MAGEA2A/MGC131923 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 4101 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| MAGEA2 | |
| MAGEA2 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human MAGEA2. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only