Learn More
MAD2L2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAD2L2.
Brand: Abnova PAB10075.100ug
Description
The protein encoded by this gene is a component of the mitotic spindle assembly checkpoint that prevents the onset of anaphase until all chromosomes are properly aligned at the metaphase plate. The encoded protein, which is similar to MAD2L1, is capable of interacting with ADAM9, ADAM15, REV1, and REV3 proteins. [provided by RefSeq
Specifications
| MAD2L2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MAD2L2. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| MAD2B/REV7 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:7500-1:30000) Western Blot (1:500-1:2000) Immunohistochemistry (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| MAD2L2 | |
| MAD2L2 | |
| A synthetic peptide corresponding to amino acids 3-14 of human MAD2L2. | |
| RUO | |
| 10459 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only