missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAD1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MAD1.
Brand: Abnova PAB12341.100uL
This item is not returnable.
View return policy
Specifications
| MAD1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MAD1. | |
| In antiserum (0.09% sodium azide) | |
| MAD1 | |
| Recombinant GST fusion protein corresponding to Yeast MAD1. | |
| RUO | |
| 852794 | |
| Yeast | |
| Liquid |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| Western Blot (1:100) The optimal working dilution should be determined by the end user. | |
| MAD1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Yeast Mad1p. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction