missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYVE1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Lyve1.
Brand: Abnova PAB12131.100uL
This item is not returnable.
View return policy
Specifications
| LYVE1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Lyve1. | |
| In Tris-glycine, 150mM NaCl (0.05% sodium azide) | |
| 1200012G08Rik/Crsbp-1/Lyve-1/Xlkd1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Flow Cytometry (1:25) The optimal working dilution should be determined by the end user. | |
| Lyve1 | |
| Lyve1 | |
| A synthetic peptide corresponding to amino acids 250-318 of mouse Lyve1. | |
| RUO | |
| 114332 | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction