Learn More
LUZP1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LUZP1.
Brand: Abnova PAB22175.100uL
Description
This gene encodes a protein that contains a leucine zipper motif. The exact function of the encoded protein is not known. In mice this gene affects neural tube closure. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: KLLASQLEEIKLQIKKQKELENGEVEGEDAFLSSKGRHERTKFRGHGSEASVSKHTARELSPQHKRERLRNREFALNNENYSLSNRQVSSPSFTSpecifications
| LUZP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant LUZP1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ35697/KIAA0601/LUZP | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 7798 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| LUZP1 | |
| LUZP1 | |
| Recombinant protein corresponding to amino acids of human LUZP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only