Learn More
LRP2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LRP2.
Brand: Abnova PAB20330.100uL
Description
The protein encoded by this gene, low density lipoprotein-related protein 2 (LRP2) or megalin, is a multi-ligand endocytic receptor that is expressed in many different tissues but primarily in absorptive epithilial tissues such as the kidney. This glycoprotein has a large amino-terminal extracellular domain, a single transmembrane domain, and a short carboxy-terminal cytoplasmic tail. The extracellular ligand-binding-domains bind diverse macromolecules including albumin, apolipoproteins B and E, and lipoprotein lipase. The LRP2 protein is critical for the reuptake of numerous ligands, including lipoproteins, sterols, vitamin-binding proteins, and hormones. This protein also has a role in cell-signaling; extracellular ligands include parathyroid horomones and the morphogen sonic hedgehog while cytosolic ligands include MAP kinase scaffold proteins and JNK interacting proteins. Recycling of this membrane receptor is regulated by phosphorylation of its cytoplasmic domain. Mutations in this gene cause Donnai-Barrow syndrome (DBS) and facio-oculoacoustico-renal syndrome (FOAR)
Sequence: GFTSMSDRPGKRCAAEGSSPLLLLPDNVRIRKYNLSSERFSEYLQDEEYIQAVDYDWDPEDIGLSVVYYTVRGEGSRFGAIKRAYIPNFESGRNNLVQEVDLKLKYVMQPDGIAVDWVGRHIYWSDVKNKRIEVAKLDGRYRKWLISTDLSpecifications
| LRP2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant LRP2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DBS/gp330 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 4036 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| LRP2 | |
| LRP2 | |
| Recombinant protein corresponding to amino acids of human LRP2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only