Learn More
LRP12, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LRP12.
Brand: Abnova PAB20931.100uL
Description
This gene was identified by its differential expression in cancer cells. The product of this gene is predicted to be a transmembrane protein. The level of this protein was found to be lower in tumor derived cell lines compared to normal cells. This gene was thus proposed to be a candidate tumor suppressor gene. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: FPVCSPNQASVLENLRLAVRSQLGFTSVRLPMAGRSSNIWNRIFNFARSRHSGSLALVSADGDEVVPSQSTSREPERNHTHRSLFSVESDDTDTENERRDMAGASGGVAAPLPQKVPPTTAVEATVGACASSSSpecifications
| LRP12 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant LRP12. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DKFZp781F1053/FLJ12929/ST7 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 29967 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| LRP12 | |
| LRP12 | |
| Recombinant protein corresponding to amino acids of human LRP12. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only