Learn More
LRMP Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LRMP.
Brand: Abnova PAB20147.100uL
Description
The protein encode dby this gene is expressed in a developmentally regulated manner in lymphoid cell lines and tissues. The protein is localized to the cytoplasmic face of the endoplasmic reticulum. [provided by RefSeq
Sequence: PSMEENGVERVCPESLLQSREYSSLPLPRHTSSTDGTITSSDPGLEILNMASCDLDRNSLCKKEEDTRSASPTIEAQGTSPAHDNIAFQDSTSKDKTILNLEAKEEPETIEEHKKEHASGDSVVSPLPVTTVKSVNVRQSENTSANEKEVSpecifications
| LRMP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant LRMP. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| JAW1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 4033 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| LRMP | |
| LRMP | |
| Recombinant protein corresponding to amino acids of human LRMP. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only