Learn More
LRG1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LRG1.
Brand: Abnova PAB28531.100uL
Description
The leucine-rich repeat (LRR) family of proteins, including LRG1, have been shown to be involved in protein-protein interaction, signal transduction, and cell adhesion and development. LRG1 is expressed during granulocyte differentiation (O'Donnell et al., 2002 [PubMed 12223515]).[supplied by OMIM
Sequence: TLSPKDCQVFRSDHGSSISCQPPAEIPGYLPADTVHLAVEFFNLTHLPANLLQGASKLQELHLSSNGLESLSPEFLRPVPQLRVLDLTRNALTGLPPGLFQASATLDTLVLKENQLEVLEVSWLHGLKALGHLDLSGSpecifications
| LRG1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant LRG1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| HMFT1766/LRG | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 116844 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:250-1:500) Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| LRG1 | |
| LRG1 | |
| Recombinant protein corresponding to amino acids of human LRG1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only