Learn More
LPIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of LPIN1.
Brand: Abnova PAB27445.100ug
Description
This gene represents a candidate gene for human lipodystrophy, characterized by loss of body fat, fatty liver, hypertriglyceridemia, and insulin resistance. Mouse studies suggest that this gene functions during normal adipose tissue development and may also play a role in human triglyceride metabolism. [provided by RefSeq
Specifications
| LPIN1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (5 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| LPIN1 | |
| LPIN1 | |
| A synthetic peptide corresponding to 18 amino acids at C-terminus of human LPIN1. | |
| 100 μg | |
| Primary | |
| LPIN1 antibody is human and mouse reactive. At least four isoforms of LPIN1 are known to exist. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of LPIN1. | |
| In PBS (0.02% sodium azide) | |
| DKFZp781P1796/KIAA0188/PAP1 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 23175 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only