Learn More
LMNB1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of LMNB1.
Brand: Abnova PAB12282.100ug
Description
The nuclear lamina consists of a two-dimensional matrix of proteins located next to the inner nuclear membrane. The lamin family of proteins make up the matrix and are highly conserved in evolution. During mitosis, the lamina matrix is reversibly disassembled as the lamin proteins are phosphorylated. Lamin proteins are thought to be involved in nuclear stability, chromatin structure and gene expression. Vertebrate lamins consist of two types, A and B. This gene encodes one of the two B type proteins, B1. [provided by RefSeq
Specifications
| LMNB1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of LMNB1. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| BC012295 | |
| LMNB1 | |
| A synthetic peptide corresponding to amino acids at C-terminus of human LMNB1. | |
| 100 μg | |
| Primary | |
| Different from the related rat and mouse sequence by three amino acids. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Immunohistochemistry (Frozen sections) (0.5-1 ug/mL) Immunocytochemistry (0.5-1 ug/mL) The optimal working dilution should be determine | |
| LMNB1 | |
| ADLD/LMN/LMN2/LMNB/MGC111419 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 4001 | |
| Human, Mouse, Rat | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only