Learn More
LMAN2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LMAN2.
Brand: Abnova PAB20249.100uL
Description
This gene encodes a type I transmembrane lectin that shuttles between the endoplasmic reticulum, the Golgi apparatus and the plasma membrane. The encoded protein binds high mannose type glycoproteins and may facilitate their sorting, trafficking and quality control. [provided by RefSeq
Sequence: GNSEHLKREHSLIKPYQGVGSSSMPLWDFQGSTMLTSQYVRLTPDERSKEGSIWNHQPCFLKDWEMHVHFKVHGTGKKNLHGDGIALWYTRDRLVPGPVFGSKDNFHGLAIFLDTYPNDETTERVFPYISVMVNNGSLSYDHSKDSpecifications
| LMAN2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant LMAN2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| C5orf8/GP36B/VIP36 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10960 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| LMAN2 | |
| LMAN2 | |
| Recombinant protein corresponding to amino acids of human LMAN2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only