missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Llama IgG (H+L) Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £206.00
Specifications
| Antigen | IgG (H+L) |
|---|---|
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunoassay |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoassay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18533512
|
Novus Biologicals
NBP1-75091 |
1 mg |
£206.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18545443
|
Novus Biologicals
NBP1-75091-0.5mg |
0.5 mg |
£155.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Llama samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunoassaySpecifications
| IgG (H+L) | |
| Western Blot, ELISA, Immunohistochemistry, Immunoassay | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Immunology | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunoassay | |
| Polyclonal | |
| Purified | |
| RUO | |
| Llama | |
| This Rabbit anti-Llama IgG (H+L) Secondary Antibody was developed against purified llama IgG (H&L). | |
| Secondary | |
| Store at 4°C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title