Learn More
LIPE Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of LIPE.
Brand: Abnova PAB12975.100ug
Description
The protein encoded by this gene has a long and a short form, generated by use of alternative translational start codons. The long form is expressed in steroidogenic tissues such as testis, where it converts cholesteryl esters to free cholesterol for steroid hormone production. The short form is expressed in adipose tissue, among others, where it hydrolyzes stored triglycerides to free fatty acids. [provided by RefSeq
Specifications
| LIPE | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of LIPE. | |
| In PBS (0.02% sodium azide) | |
| HSL/LHS | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| LIPE | |
| LIPE | |
| A synthetic peptide corresponding to C-terminus 14 amino acids of human LIPE. | |
| RUO | |
| 3991 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only