Learn More
LGALS4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of LGALS4.
Brand: Abnova PAB24962.100uL
Description
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The expression of this gene is restricted to small intestine, colon, and rectum, and it is underexpressed in colorectal cancer. [provided by RefSeq
Specifications
| LGALS4 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| LGALS4 | |
| LGALS4 | |
| A synthetic peptide corresponding to LGALS4. | |
| 100 μL | |
| Primary | |
| LGALS4 polyclonal antibody detects endogenous levels of LGALS4 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of LGALS4. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| GAL4/L36LBP | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3960 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only