Learn More
LGALS2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LGALS2.
Brand: Abnova PAB20221.100uL
Description
The protein encoded by this gene is a soluble beta-galactoside binding lectin. The encoded protein is found as a homodimer and can bind to lymphotoxin-alpha. A single nucleotide polymorphism in an intron of this gene can alter the transcriptional level of the protein, with a resultant increased risk of myocardial infarction. [provided by RefSeq
Sequence: LEVKNMDMKPGSTLKITGSIADGTDGFVINLGQGTDKLNLHFNPRFSESTIVCNSLDGSNWGQEQREDHLCFSPGSEVKFTVTFESDKFKVKLPDGHELTFPNRLGHSHLSYLSVRGGFNMSSFKLSpecifications
| LGALS2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant LGALS2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| HL14/MGC75071 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3957 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| LGALS2 | |
| LGALS2 | |
| Recombinant protein corresponding to amino acids of human LGALS2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only