Learn More
LEP Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant of LEP.
Brand: Abnova PAB5093.150ug
Description
This gene encodes a protein that is secreted by white adipocytes, and which plays a major role in the regulation of body weight. This protein, which acts through the leptin receptor, functions as part of a signaling pathway that can inhibit food intake and/or regulate energy expenditure to maintain constancy of the adipose mass. This protein also has several endocrine functions, and is involved in the regulation of immune and inflammatory responses, hematopoiesis, angiogenesis and wound healing. Mutations in this gene and/or its regulatory regions cause severe obesity, and morbid obesity with hypogonadism. This gene has also been linked to type 2 diabetes mellitus development. [provided by RefSeq
Specifications
| LEP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant of LEP. | |
| In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal) | |
| FLJ94114/OB/OBS | |
| Rabbit | |
| 150 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| LEP | |
| LEP | |
| Recombinant protein corresponding to human LEP. | |
| RUO | |
| 3952 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only