Learn More
LEO1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LEO1.
Brand: Abnova PAB23688.100uL
Description
LEO1, parafibromin (CDC73; MIM 607393), CTR9 (MIM 609366), and PAF1 (MIM 610506) form the PAF protein complex that associates with the RNA polymerase II subunit POLR2A (MIM 180660) and with a histone methyltransferase complex (Rozenblatt-Rosen et al., 2005 [PubMed 15632063]).[supplied by OMIM
Sequence: SGQPSNKELFGDDSEDEGASHHSGSDNHSERSDNRSEASERSDHEDNDPSDVDQHSGSEAPNDDEDEGHRSDGGSHHSEAEGSEKAHSDDEKWGREDKSDQSDDEKIQNSDSpecifications
| LEO1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant LEO1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| RDL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 123169 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| LEO1 | |
| LEO1 | |
| Recombinant protein corresponding to amino acids of human LEO1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only