missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEMD1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of LEMD1.
Brand: Abnova PAB25769.100ug
This item is not returnable.
View return policy
Specifications
| LEMD1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| LEMD1 | |
| LEMD1 | |
| A synthetic peptide corresponding to 19 amino acids at internal region of human LEMD1. | |
| 100 μg | |
| Primary | |
| At least six isoforms of LEMD1 are known to exist; this antibody will detect the longest isoform. LEMD1 antibody is predicted to not cross-react with LEMD2 and LEMD3. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of LEMD1. | |
| In PBS (0.02% sodium azide) | |
| LEMP-1 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 93273 | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction