Learn More
LDHC, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LDHC.
Brand: Abnova PAB28316.100uL
Description
Lactate dehydrogenase C catalyzes the conversion of L-lactate and NAD to pyruvate and NADH in the final step of anaerobic glycolysis. LDHC is testis-specific and belongs to the lactate dehydrogenase family. Two transcript variants have been detected which differ in the 5' untranslated region. [provided by RefSeq]
Sequence: MSTVKEQLIEKLIEDDENSQCKITIVGTGAVGSpecifications
| LDHC | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant LDHC. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| CT32/LDH3/LDHX/MGC111073 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3948 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Immunohistochemistry (Formalin/Paraffin), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:2500-1:5000) The optimal working dilution should be determined by the end user. | |
| LDHC | |
| LDHC | |
| Recombinant protein corresponding to amino acids of recombinant LDHC. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only