missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lbr, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Lbr.
Brand: Abnova PAB15583.50ug
This item is not returnable.
View return policy
Specifications
| Lbr | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant Lbr. | |
| In antiserum (0.05% sodium azide) | |
| AI505894/ic | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Lbr | |
| Lbr | |
| Recombinant protein corresponding to amino acids 1-81 mouse Lbr. | |
| RUO | |
| 98386 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction