Learn More
LARS Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of LARS.
Brand: Abnova PAB2886.400uL
Description
This gene encodes a cytosolic leucine-tRNA synthetase, a member of the class I aminoacyl-tRNA synthetase family. The encoded enzyme catalyzes the ATP-dependent ligation of L-leucine to tRNA(Leu). It is found in the cytoplasm as part of a multisynthetase complex and interacts with the arginine tRNA synthetase through its C-terminal domain. Alternatively spliced transcript variants of this gene have been found; however, their full-length nature is not known. [provided by RefSeq
Specifications
| LARS | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of LARS. | |
| In PBS (0.09% sodium azide) | |
| FLJ10595/FLJ21788/HSPC192/KIAA1352/LARS1/LEURS/LEUS/LRS/PIG44/RNTLS/hr025Cl | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 51520 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemsitry (1:10-50) The optimal working dilution should be determined by the end user. | |
| LARS | |
| LARS | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human LARS. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only