Learn More
KTN1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant KTN1.
Brand: Abnova PAB20173.100uL
Description
Various cellular organelles and vesicles are transported along the microtubules in the cytoplasm. Likewise, membrane recycling of the endoplasmic reticulum (ER), Golgi assembly at the microtubule organizing center, and alignment of lysosomes along microtubules are all related processes. The transport of organelles requires a special class of microtubule-associated proteins (MAPs). One of these is the molecular motor kinesin (see MIM 148760 and MIM 600025), an ATPase that moves vesicles unidirectionally toward the plus end of the microtubule. Another such MAP is kinectin, a large integral ER membrane protein. Antibodies directed against kinectin have been shown to inhibit its binding to kinesin.[supplied by OMIM
Sequence: ETLMVPSKRQEALPLHQETKQESGSGKKKASSKKQKTENVFVDEPLIHATTYIPLMDNADSSPVVDKREVIDLLKPDQVEGIQKSGTKKLKTETDKENAEVKFKDFLLSLKTMMFSEDEALCVVDLLKEKSGVIQSpecifications
| KTN1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant KTN1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| CG1/KIAA0004/KNT/MGC133337/MU-RMS-40.19 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3895 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| KTN1 | |
| KTN1 | |
| Recombinant protein corresponding to amino acids of human KTN1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only