missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KTI3 (Lima bean), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native KTI3 (Lima bean).
Brand: Abnova PAB28090.100ug
This item is not returnable.
View return policy
Specifications
| KTI3 (Lima bean) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native KTI3 (Lima bean). | |
| Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| KTi | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Plant | |
| Lyophilized |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:5000 - 1:25000) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user. | |
| KTi3 | |
| KTi3 | |
| Native purified Lima bean KTI3. | |
| RUO | |
| 547831 | |
| Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction