Learn More
KREMEN2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KREMEN2.
Brand: Abnova PAB27402.100ug
Description
This gene encodes a high-affinity dickkopf homolog 1 (DKK1) transmembrane receptor that functionally cooperates with DKK1 to block wingless (WNT)/beta-catenin signaling. The encoded protein forms a ternary membrane complex with DKK1 and the WNT receptor lipoprotein receptor-related protein 6 (LRP6), and induces rapid endocytosis and removal of LRP6 from the plasma membrane. It contains extracellular kringle, WSC, and CUB domains. Alternatively spliced transcript variants encoding distinct isoforms have been observed for this gene. [provided by RefSeq
Specifications
| KREMEN2 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunocytochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| KREMEN2 | |
| KREMEN2 | |
| A synthetic peptide corresponding to 15 amino acids at C-terminus of human KREMEN2. | |
| 100 μg | |
| Primary | |
| Multiple isoforms of KREMEN2 are known to exist. KREMEN2 antibody is predicted to not cross-react with other Kremen protein family members. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of KREMEN2. | |
| In PBS (0.02% sodium azide) | |
| KRM2/MGC10791/MGC16709 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 79412 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only