Learn More
KREMEN1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KREMEN1.
Brand: Abnova PAB27396.100ug
Description
This gene encodes a high-affinity dickkopf homolog 1 (DKK1) transmembrane receptor that functionally cooperates with DKK1 to block wingless (WNT)/beta-catenin signaling. The encoded protein is a component of a membrane complex that modulates canonical WNT signaling through lipoprotein receptor-related protein 6 (LRP6). It contains extracellular kringle, WSC, and CUB domains. Alternatively spliced transcript variants encoding distinct isoforms have been observed for this gene. [provided by RefSeq
Specifications
| KREMEN1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) Immunohistochemistry (5 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| KREMEN1 | |
| KREMEN1 | |
| A synthetic peptide corresponding to 18 amino acids at C-terminus of human KREMEN1. | |
| 100 μg | |
| Primary | |
| Three isoforms of KREMEN1 exists as a result of alternative splicing event. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of KREMEN1. | |
| In PBS (0.02% sodium azide) | |
| FLJ31863/KREMEM1/KREMEN/KRM1 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 83999 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only