missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KPNA7 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KPNA7.
Brand: Abnova PAB27189.100ug
This item is not returnable.
View return policy
Specifications
| KPNA7 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| KPNA7 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 402569 | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of KPNA7. | |
| In PBS (0.02% sodium azide) | |
| KPNA7 | |
| A synthetic peptide corresponding to 13 amino acids at N-terminus of human KPNA7. | |
| 100 μg | |
| Primary | |
| KPNA7 antibody is human specific. KPNA7 antibody is predicted to not cross-react with other Importin alpha family members. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction