Learn More
KLK4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KLK4.
Brand: Abnova PAB1994.400uL
Description
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. In some tissues its expression is hormonally regulated. The expression pattern of a similar mouse protein in murine developing teeth supports a role for the protein in the degradation of enamel proteins. Alternate splice variants for this gene have been described, but their biological validity has not been determined. [provided by RefSeq
Specifications
| KLK4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of KLK4. | |
| In PBS (0.09% sodium azide) | |
| ARM1/EMSP/EMSP1/KLK-L1/MGC116827/MGC116828/PROSTASE/PRSS17/PSTS | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 9622 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| KLK4 | |
| KLK4 | |
| A synthetic peptide (conjugated with KLH) corresponding to internal region of human KLK4. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only