missing translation for 'onlineSavingsMsg'
Learn More

Klhl29, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against partial recombinant Klhl29.

Brand:  Abnova PAB15855.50ug

Product Code. 16071847

  • £535.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Klhl29
Polyclonal
Rabbit polyclonal antibody raised against partial recombinant Klhl29.
In PBS (50% glycerol, 0.02% sodium azide)
A230106N14Rik/Gm68/Kbtbd9/mKIAA1921
Rabbit
50 μg
Primary
Specific to recombinant protein GX0065. This antibody detects mKLHL29 protein.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Western Blot
Unconjugated
Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Klhl29
Klhl29
Recombinant GST fusion protein corresponding to 278 amino acids of mouse Klhl29.
RUO
208439
Mouse
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only